The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009941.2 | NP_001009941.1 | 4 - 5 | 92 | |
NM_001009943.2 | NP_001009943.1 | 4 - 5 | 92 | ||
NM_019046.2 | NP_061919.1 | 4 - 5 | 92 | ||
GenBank mRNA | AI738771.1 | - | 2 - 3 | 362 | 92 |
AL137614.1 | - | 1 - 2 | 92 | ||
AW470888.1 | - | 3 - 4 | 338 | 92 | |
BC062346.1 | - | 4 - 5 | 92 |