Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT5DC4
Entrez Gene ID
284958
Gene Name
5'-nucleotidase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.2: 112717245 - 112742879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011512262.2 XP_011510564.1 1 - 2 404 66
XM_017005474.1 XP_016860963.1 1 - 2 404 66
XM_017005475.1 XP_016860964.1 1 - 2 404 66
XM_017005476.1 XP_016860965.1 2 - 3 419 66
XM_017005477.1 XP_016860966.1 2 - 3 2014 66
XM_017005478.1 XP_016860967.1 2 - 3 145 66
GenBank mRNA BC041437.1 - 2 - 3 137 66

View Full Product Details