Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MEG8
- Entrez Gene ID
-
79104
- Gene Name
- maternally expressed 8 (non-protein coding)
- Gene Aliases
-
Bsr, Irm, LINC00024, NCRNA00024, Rian
- Chromosome Location
-
Chr.14: 100894770 - 100906968 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024149.2 |
|
1 - 2 |
270 |
65 |
GenBank mRNA |
CA396130.1 |
- |
1 - 2 |
273 |
65 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
T85042.1 |
- |
|
|
|
Chr. - - [-] on Build |
64 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, C/D box 112 |
SNORD112 |
14q(0) |
|
Sense |
Chr. - 100897920 - 100897996 on Build GRCh38 |
Downstream |
microRNA 370 |
MIR370 |
MIRN370, hsa-mir-370, mir-370 |
12.1kb
|
Sense |
Chr. - 100911139 - 100911213 on Build GRCh38 |
View Full Product Details