The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164462.1 | NP_001157934.1 | 5 - 6 | 15247 | 88 |
GenBank mRNA | AF147790.1 | - | 4 - 5 | 999 | 88 |
AK024856.1 | - | 4 - 5 | 754 | 88 | |
BC127877.1 | - | 4 - 5 | 1017 | 88 |