The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015306.2 | NP_056121.2 | 18 - 19 | 2368 | 118 |
XM_005270690.3 | XP_005270747.1 | 5 - 6 | 409 | 118 | |
XM_017000831.1 | XP_016856320.1 | 19 - 20 | 2433 | 118 | |
XM_017000832.1 | XP_016856321.1 | 19 - 20 | 2430 | 118 | |
XM_017000833.1 | XP_016856322.1 | 19 - 20 | 1925 | 118 | |
XM_017000834.1 | XP_016856323.1 | 18 - 19 | 1865 | 118 | |
XM_017000835.1 | XP_016856324.1 | 19 - 20 | 2436 | 118 | |
XM_017000836.1 | XP_016856325.1 | 19 - 20 | 2436 | 118 | |
XM_017000837.1 | XP_016856326.1 | 19 - 20 | 2436 | 118 | |
GenBank mRNA | AK127075.1 | - | 1 - 2 | 113 | 118 |