Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP53
Entrez Gene ID
54532
Gene Name
ubiquitin specific peptidase 53
Gene Aliases
-
Chromosome Location
Chr.4: 119212583 - 119295518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011532037.2 XP_011530339.1 1 - 2 297 88

View Full Product Details