The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001071.2 | NP_001062.1 | 5 - 6 | 874 | 69 |
GenBank mRNA | AB062290.1 | - | 6 - 7 | 816 | 69 |
AB077207.1 | - | 3 - 4 | 486 | 69 | |
AB077208.1 | - | 4 - 5 | 633 | 69 | |
AK223372.1 | - | 5 - 6 | 862 | 69 | |
AK292620.1 | - | 5 - 6 | 769 | 69 | |
AK314140.1 | - | 5 - 6 | 833 | 69 | |
BC002567.2 | - | 5 - 6 | 803 | 69 | |
BC013919.1 | - | 6 - 7 | 830 | 69 | |
BC018858.2 | - | 5 - 6 | 787 | 69 | |
BC083512.1 | - | 6 - 7 | 841 | 69 | |
BT006811.1 | - | 5 - 6 | 735 | 69 | |
X02308.1 | - | 5 - 6 | 840 | 69 |