Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEFA6
Entrez Gene ID
1671
Gene Name
defensin alpha 6
Gene Aliases
DEF6, HD-6
Chromosome Location
Chr.8: 6924694 - 6926076 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001926.3 NP_001917.1 1 - 2 238 65
GenBank mRNA BC069667.1 - 1 - 2 238 65
BC069710.1 - 1 - 2 238 65
BC069728.1 - 1 - 2 238 65
BC069769.1 - 1 - 2 240 65
BC093951.1 - 1 - 2 218 65
BC093953.1 - 1 - 2 218 65
M98331.1 - 1 - 2 215 65

View Full Product Details