The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003837.3 | NP_003828.2 | 6 - 7 | 932 | 65 |
GenBank mRNA | BC113632.1 | - | 6 - 7 | 862 | 65 |
BC117477.1 | - | 6 - 7 | 862 | 65 | |
CR536483.1 | - | 6 - 7 | 826 | 65 | |
Y10812.2 | - | 6 - 7 | 893 | 65 |