The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003860.3 | NP_003851.1 | 1 - 2 | 490 | 111 |
GenBank mRNA | AF068235.1 | - | 1 - 2 | 489 | 110 |
BG748852.1 | - | 1 - 2 | 273 | 111 |