The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018108.3 | NP_001018118.1 | 2 - 3 | 648 | 69 |
NM_001199877.1 | NP_001186806.1 | 4 - 5 | 1046 | 69 | |
NM_001199878.1 | NP_001186807.1 | 2 - 3 | 341 | 69 | |
NR_037672.1 | 2 - 3 | 514 | 69 | ||
GenBank mRNA | AF073298.1 | - | 2 - 3 | 172 | 69 |
AF320073.1 | - | 2 - 3 | 166 | 69 | |
AK000479.1 | - | 2 - 3 | 160 | 69 | |
AL049976.1 | - | 2 - 3 | 185 | 69 | |
AV713454.2 | - | 2 - 3 | 211 | 69 | |
BC008214.1 | - | 2 - 3 | 648 | 69 | |
BC009869.2 | - | 4 - 5 | 1044 | 69 | |
BC015491.1 | - | 2 - 3 | 145 | 69 | |
BC022326.1 | - | 2 - 3 | 163 | 69 | |
BC038893.1 | - | 2 - 3 | 129 | 69 | |
BI260545.1 | - | 2 - 3 | 248 | 69 | |
DA302996.1 | - | 2 - 3 | 342 | 69 | |
DB060650.1 | - | 2 - 3 | 170 | 69 |