The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014849.4 | NP_055664.3 | 2 - 3 | 1118 | 59 |
XM_017002992.1 | XP_016858481.1 | 2 - 3 | 904 | 59 | |
GenBank mRNA | AB018279.1 | - | 2 - 3 | 1061 | 59 |
AK075480.1 | - | 2 - 3 | 1078 | 59 | |
AK291718.1 | - | 2 - 3 | 1067 | 59 | |
BC000776.2 | - | 1 - 2 | 213 | 59 | |
BC045111.1 | - | 2 - 3 | 1064 | 59 | |
BX537754.1 | - | 2 - 3 | 1114 | 59 |