The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014849.4 | NP_055664.3 | 7 - 8 | 1782 | 62 |
XM_017002992.1 | XP_016858481.1 | 7 - 8 | 1568 | 62 | |
GenBank mRNA | AB018279.1 | - | 7 - 8 | 1725 | 62 |
AK075480.1 | - | 7 - 8 | 1742 | 62 | |
AK291718.1 | - | 7 - 8 | 1731 | 62 | |
BC000776.2 | - | 6 - 7 | 877 | 62 | |
BC045111.1 | - | 7 - 8 | 1728 | 62 | |
BX537754.1 | - | 7 - 8 | 1778 | 62 |