Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEMO1
Entrez Gene ID
51072
Gene Name
mediator of cell motility 1
Gene Aliases
C2orf4, CGI-27, MEMO, NS5ATP7
Chromosome Location
Chr.2: 31867188 - 32011052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137602.2 NP_001131074.1 7 - 8 1106 103
NM_001301833.1 NP_001288762.1 9 - 10 886 103
NM_001301852.1 NP_001288781.1 7 - 8 665 103
NM_015955.3 NP_057039.1 8 - 9 1175 103
NR_126032.1 6 - 7 955 103
NR_126034.1 6 - 7 588 103
XM_011532892.2 XP_011531194.1 9 - 10 823 103
XM_011532896.2 XP_011531198.1 6 - 7 558 103
XM_017004255.1 XP_016859744.1 9 - 10 796 103
GenBank mRNA AB041018.1 - 8 - 9 1175 103
AB041019.1 - 8 - 9 824 103
AF132961.1 - 8 - 9 873 103
AF363446.1 - 8 - 9 1092 103
AF529368.1 - 8 - 9 758 103
AK057760.1 - 6 - 7 577 103
AK290753.1 - 9 - 10 842 103
AK297128.1 - 7 - 8 817 103
AL161966.1 - 2 - 2 147 103
BC018733.1 - 8 - 9 1140 103
BC070046.1 - 9 - 10 915 103
BC094681.1 - 8 - 9 779 103
CD250945.1 - 6 - 7 652 103

View Full Product Details