The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184998.1 | NP_001171927.1 | 18 - 19 | 3818 | 73 |
NM_001184999.1 | NP_001171928.1 | 18 - 19 | 3809 | 73 | |
NM_014647.3 | NP_055462.2 | 18 - 19 | 3818 | 73 | |
XM_005255704.1 | XP_005255761.1 | 18 - 19 | 3814 | 73 | |
XM_005255706.1 | XP_005255763.1 | 18 - 19 | 3808 | 73 | |
XM_005255707.1 | XP_005255764.1 | 18 - 19 | 3784 | 73 | |
XM_017023900.1 | XP_016879389.1 | 18 - 19 | 3702 | 73 | |
XM_017023901.1 | XP_016879390.1 | 18 - 19 | 3699 | 73 | |
XM_017023902.1 | XP_016879391.1 | 17 - 18 | 3652 | 73 | |
XM_017023903.1 | XP_016879392.1 | 17 - 18 | 3385 | 73 | |
XM_017023904.1 | XP_016879393.1 | 17 - 18 | 3382 | 73 | |
XM_017023905.1 | XP_016879394.1 | 18 - 19 | 3239 | 73 | |
XM_017023906.1 | XP_016879395.1 | 18 - 19 | 3180 | 73 | |
GenBank mRNA | AB007890.3 | - | 16 - 17 | 2904 | 73 |
BC137165.1 | - | 18 - 19 | 3760 | 73 | |
BC137170.1 | - | 18 - 19 | 3760 | 73 | |
BC144514.1 | - | 18 - 19 | 3760 | 73 | |
BC144515.1 | - | 18 - 19 | 3751 | 73 |