Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF20
Entrez Gene ID
79853
Gene Name
transmembrane 4 L six family member 20
Gene Aliases
PRO994, SLI5, TCCE518
Chromosome Location
Chr.2: 227362041 - 227379306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024795.3 NP_079071.2 1 - 2 226 92
GenBank mRNA AK026453.1 - 1 - 2 225 92
AY358671.1 - 1 - 2 218 92
BC035754.1 - 1 - 2 213 92
BC137256.1 - 1 - 2 212 92
BC137257.1 - 1 - 2 212 92
DA926038.1 - 1 - 2 226 92

View Full Product Details