Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236467740-236468325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDARADD
Entrez Gene ID
128178
Gene Name
EDAR associated death domain
Gene Aliases
ECTD11A, ECTD11B, ED3, EDA3
UniGene
Hs.352224
Gene Chromosome Location
Chr.1: 236394380-236484708 on Build GRCh38
Transcripts
NM_080738.3 NM_145861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651549 c.226G>C p.E76Q Chr.1: 236468237-236468237 on Build GRCh38
5845932 c.233C>T p.P78L Chr.1: 236468274-236468274 on Build GRCh38
6062127 c.246C>T p.S82S Chr.1: 236468257-236468257 on Build GRCh38
6651550 c.196G>C p.E66Q Chr.1: 236468237-236468237 on Build GRCh38
6062128 c.216C>T p.S72S Chr.1: 236468257-236468257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376772514 Chr.1: 236468265-236468265 on Build GRCh38
rs968813348 Chr.1: 236468144-236468144 on Build GRCh38
rs1013860556 Chr.1: 236467990-236467990 on Build GRCh38
rs751288720 Chr.1: 236467784-236467784 on Build GRCh38
rs540478996 Chr.1: 236468142-236468142 on Build GRCh38
rs140420342 Chr.1: 236467979-236467979 on Build GRCh38
rs35734689 Chr.1: 236468100-236468101 on Build GRCh38
rs544791748 Chr.1: 236467765-236467765 on Build GRCh38
rs1048265998 Chr.1: 236468047-236468047 on Build GRCh38
rs944718430 Chr.1: 236468073-236468073 on Build GRCh38
rs187389302 Chr.1: 236468054-236468054 on Build GRCh38
rs1253601 Chr.1: 236467791-236467791 on Build GRCh38
rs372150890 Chr.1: 236468199-236468199 on Build GRCh38
rs570899265 Chr.1: 236467949-236467949 on Build GRCh38
rs138967527 Chr.1: 236468269-236468269 on Build GRCh38
rs998516716 Chr.1: 236468157-236468157 on Build GRCh38
rs904060537 Chr.1: 236468244-236468244 on Build GRCh38
rs888416016 Chr.1: 236467862-236467862 on Build GRCh38
rs768049797 Chr.1: 236468279-236468279 on Build GRCh38
rs143660241 Chr.1: 236467880-236467880 on Build GRCh38
rs201611000 Chr.1: 236468299-236468299 on Build GRCh38
rs150720000 Chr.1: 236468193-236468193 on Build GRCh38
rs947127322 Chr.1: 236467765-236467767 on Build GRCh38
rs1033866648 Chr.1: 236468118-236468118 on Build GRCh38
rs530374757 Chr.1: 236467806-236467806 on Build GRCh38
rs769609962 Chr.1: 236467765-236467765 on Build GRCh38
rs554873612 Chr.1: 236468045-236468045 on Build GRCh38
rs776471811 Chr.1: 236468223-236468223 on Build GRCh38
rs932540075 Chr.1: 236467834-236467834 on Build GRCh38
rs548947836 Chr.1: 236467822-236467822 on Build GRCh38
rs1027457554 Chr.1: 236468039-236468039 on Build GRCh38
rs1033961467 Chr.1: 236467939-236467939 on Build GRCh38
rs906733936 Chr.1: 236467894-236467894 on Build GRCh38
rs968404871 Chr.1: 236467997-236467997 on Build GRCh38
rs568749836 Chr.1: 236468019-236468019 on Build GRCh38
rs112248042 Chr.1: 236467947-236467947 on Build GRCh38
rs943883523 Chr.1: 236467793-236467793 on Build GRCh38
rs769534560 Chr.1: 236468227-236468227 on Build GRCh38
rs1024029913 Chr.1: 236468014-236468014 on Build GRCh38
rs892540789 Chr.1: 236467847-236467847 on Build GRCh38
rs977613068 Chr.1: 236468001-236468001 on Build GRCh38
rs1045342641 Chr.1: 236468284-236468284 on Build GRCh38
rs78452294 Chr.1: 236468052-236468052 on Build GRCh38
rs1037547189 Chr.1: 236468077-236468077 on Build GRCh38
rs747953067 Chr.1: 236468224-236468224 on Build GRCh38
rs906605643 Chr.1: 236468078-236468078 on Build GRCh38
rs1044285599 Chr.1: 236468231-236468231 on Build GRCh38
rs895281473 Chr.1: 236468134-236468134 on Build GRCh38
rs763211638 Chr.1: 236468253-236468253 on Build GRCh38
rs768595907 Chr.1: 236468212-236468212 on Build GRCh38
rs897579878 Chr.1: 236468296-236468296 on Build GRCh38
rs1055330533 Chr.1: 236468260-236468260 on Build GRCh38
rs965116592 Chr.1: 236468185-236468185 on Build GRCh38
rs766731640 Chr.1: 236468257-236468257 on Build GRCh38
rs142730964 Chr.1: 236468040-236468040 on Build GRCh38
rs1048399033 Chr.1: 236467837-236467837 on Build GRCh38
rs1011195768 Chr.1: 236468138-236468138 on Build GRCh38
rs1015673696 Chr.1: 236467888-236467888 on Build GRCh38
rs778788277 Chr.1: 236468217-236468217 on Build GRCh38
rs886046185 Chr.1: 236468216-236468216 on Build GRCh38
rs556698868 Chr.1: 236468010-236468010 on Build GRCh38
rs1031723003 Chr.1: 236468172-236468174 on Build GRCh38
rs758046261 Chr.1: 236468188-236468188 on Build GRCh38
rs1003601106 Chr.1: 236468094-236468094 on Build GRCh38
rs1043877993 Chr.1: 236467861-236467861 on Build GRCh38
rs370659008 Chr.1: 236468169-236468169 on Build GRCh38
rs776222825 Chr.1: 236467977-236467977 on Build GRCh38
rs773065240 Chr.1: 236468242-236468242 on Build GRCh38
rs972319125 Chr.1: 236468035-236468035 on Build GRCh38
rs367622669 Chr.1: 236468285-236468285 on Build GRCh38
rs527719045 Chr.1: 236467904-236467904 on Build GRCh38
rs117338854 Chr.1: 236468053-236468053 on Build GRCh38
rs4565703 Chr.1: 236468291-236468291 on Build GRCh38
rs945561273 Chr.1: 236467859-236467859 on Build GRCh38
rs1033890578 Chr.1: 236467919-236467919 on Build GRCh38

View Full Product Details