The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001547.4 | NP_001538.4 | 1 - 2 | 176 | 66 |
GenBank mRNA | AK300431.1 | - | 1 - 2 | 78 | 66 |
AK312831.1 | - | 1 - 2 | 125 | 66 | |
BC005987.1 | - | 1 - 2 | 79 | 66 | |
BC032839.2 | - | 1 - 2 | 79 | 66 | |
DA631671.1 | - | 1 - 2 | 180 | 66 |