The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006735.3 | NP_006726.1 | 1 - 2 | 670 | 107 |
GenBank mRNA | AW292406.1 | - | 1 - 2 | 186 | 107 |
BC130571.1 | - | 1 - 2 | 424 | 107 | |
BC136500.1 | - | 1 - 2 | 424 | 107 | |
BE465040.1 | - | 1 - 2 | 135 | 107 |