The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017669.2 | NP_060139.2 | 2 - 2 | 2356 | 141 |
GenBank mRNA | AK000112.1 | - | 1 - 1 | 453 | 141 |
AK074719.1 | - | 1 - 1 | 1830 | 141 | |
AY121802.1 | - | 1 - 1 | 453 | 141 | |
BC111486.1 | - | 2 - 2 | 2279 | 141 | |
EU069463.1 | - | 2 - 2 | 2259 | 141 |