Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INF2
Entrez Gene ID
64423
Gene Name
inverted formin, FH2 and WH2 domain containing
Gene Aliases
C14orf151, C14orf173, CMTDIE, FSGS5, pp9484
Chromosome Location
Chr.14: 104689606 - 104719610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031714.3 NP_001026884.3 2 - 3 537 68
NM_022489.3 NP_071934.3 2 - 3 537 68
NM_032714.2 NP_116103.1 2 - 3 537 68
XM_005268004.4 XP_005268061.1 2 - 3 624 68
XM_005268005.4 XP_005268062.1 2 - 3 595 68
XM_017021595.1 XP_016877084.1 2 - 3 595 68
GenBank mRNA AK290083.1 - 2 - 3 522 68
BC006173.1 - 2 - 3 522 68
BI489810.1 - 2 - 3 537 68
BX248757.1 - 2 - 3 506 68

View Full Product Details