Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19orf18
Entrez Gene ID
147685
Gene Name
chromosome 19 open reading frame 18
Gene Aliases
-
Chromosome Location
Chr.19: 57958437 - 57974534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152474.4 NP_689687.1 4 - 5 478 73
GenBank mRNA BC033933.2 - 4 - 5 481 73

View Full Product Details