Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
Chromosome Location
Chr.X: 101418263 - 101535988 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028407.2 6 - 7 499 64
NR_045861.1 3 - 4 426 64
NR_045862.1 3 - 4 432 64
NR_045863.1 3 - 4 426 64
NR_045864.1 3 - 4 432 64
GenBank mRNA AK096955.1 - 3 - 4 224 64
AK125041.1 - 3 - 4 230 64
AK126987.1 - 3 - 4 253 64
AK292543.1 - 3 - 4 259 64
BC032236.1 - 3 - 4 205 64
BC036206.1 - 3 - 4 428 64
CR933662.1 - 6 - 7 474 64
DA450941.1 - 6 - 7 486 64
DA531702.1 - 3 - 4 250 64

View Full Product Details