The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319953.1 | NP_001306882.1 | 2 - 3 | 462 | 81 |
NM_153007.4 | NP_694552.2 | 2 - 3 | 777 | 81 | |
XM_005256476.3 | XP_005256533.1 | 3 - 4 | 630 | 81 | |
XM_011523688.1 | XP_011521990.1 | 3 - 4 | 399 | 81 | |
GenBank mRNA | AB081120.1 | - | 2 - 3 | 777 | 81 |
AY237799.1 | - | 2 - 3 | 777 | 81 | |
BC112010.1 | - | 2 - 3 | 691 | 81 | |
BC112014.1 | - | 2 - 3 | 691 | 81 | |
FJ235526.1 | - | 2 - 3 | 432 | 81 |