Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP50
Entrez Gene ID
10762
Gene Name
nucleoporin 50
Gene Aliases
NPAP60, NPAP60L
Chromosome Location
Chr.22: 45163845 - 45188011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153645.2 NP_705931.1 2 - 3 165 132
XM_005261312.1 XP_005261369.1 2 - 3 389 132
XM_005261314.1 XP_005261371.1 2 - 3 389 132
XM_006724104.1 XP_006724167.1 2 - 3 199 132
XM_017028537.1 XP_016884026.1 2 - 3 329 132
GenBank mRNA AL704529.1 - 2 - 3 132
BC016055.1 - 2 - 3 308 132
BC028125.1 - 2 - 3 165 132

View Full Product Details