The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153705.4 | NP_714916.3 | 2 - 3 | 460 | 71 |
XM_011542621.2 | XP_011540923.1 | 2 - 3 | 433 | 71 | |
XM_011542623.2 | XP_011540925.1 | 2 - 3 | 427 | 71 | |
GenBank mRNA | AK125519.1 | - | 1 - 2 | 242 | 71 |
AK313534.1 | - | 2 - 3 | 322 | 71 | |
AY358616.1 | - | 1 - 2 | 529 | 71 | |
BC036526.2 | - | 2 - 3 | 414 | 71 |