The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002617.3 | NP_002608.1 | 1 - 2 | 183 | 112 |
NM_153818.1 | NP_722540.1 | 1 - 2 | 183 | 112 | |
XM_011541573.1 | XP_011539875.1 | 1 - 2 | 234 | 112 | |
XM_011541576.1 | XP_011539878.1 | 1 - 2 | 234 | 112 | |
GenBank mRNA | AB013818.1 | - | 1 - 2 | 183 | 112 |
AF060502.1 | - | 1 - 2 | 146 | 112 | |
AK124816.1 | - | 1 - 2 | 161 | 112 | |
BC000543.2 | - | 1 - 2 | 142 | 112 | |
BC018198.1 | - | 1 - 2 | 165 | 112 | |
CR542077.1 | - | 1 - 2 | 114 | 112 | |
DA439481.1 | - | 1 - 2 | 184 | 113 |