The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002617.3 | NP_002608.1 | 4 - 5 | 847 | 50 |
NM_153818.1 | NP_722540.1 | 4 - 5 | 907 | 50 | |
XM_011541573.1 | XP_011539875.1 | 4 - 5 | 955 | 50 | |
GenBank mRNA | AB013818.1 | - | 4 - 5 | 847 | 50 |
AF060502.1 | - | 4 - 5 | 810 | 50 | |
AK124816.1 | - | 4 - 5 | 825 | 50 | |
BC000543.2 | - | 4 - 5 | 866 | 50 | |
BC018198.1 | - | 4 - 5 | 829 | 50 | |
CR542077.1 | - | 4 - 5 | 778 | 50 |