Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYRIP
Entrez Gene ID
25924
Gene Name
myosin VIIA and Rab interacting protein
Gene Aliases
SLAC2-C, SLAC2C
Chromosome Location
Chr.3: 39808914 - 40260321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284423.1 NP_001271352.1 12 - 13 2204 65
NM_001284425.1 NP_001271354.1 11 - 12 2256 65
NM_001284426.1 NP_001271355.1 10 - 11 2014 65
NM_015460.3 NP_056275.2 12 - 13 2393 65
NR_104316.1 12 - 13 2356 65
XM_011533575.1 XP_011531877.1 12 - 13 2365 65
GenBank mRNA AB083783.1 - 11 - 12 2106 65
AF396687.1 - 12 - 13 2242 65
AK125334.1 - 10 - 11 1880 65
AK294714.1 - 10 - 11 2004 65
AK312412.1 - 12 - 13 2203 65
AK316068.1 - 10 - 11 2014 65
BC092512.1 - 12 - 13 2392 65
BC109311.1 - 11 - 12 2084 65
BC109312.1 - 12 - 13 2184 65

View Full Product Details