The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286455.1 | NP_001273384.1 | 10 - 10 | 2729 | 80 |
NM_015482.1 | NP_056297.1 | 10 - 10 | 3188 | 80 | |
NM_021945.6 | NP_068764.3 | 11 - 11 | 2974 | 80 | |
NR_104448.1 | 12 - 12 | 2445 | 80 | ||
XM_011514801.2 | XP_011513103.1 | 11 - 11 | 3275 | 80 | |
XM_017011180.1 | XP_016866669.1 | 11 - 11 | 3752 | 80 | |
XM_017011181.1 | XP_016866670.1 | 9 - 9 | 2501 | 80 | |
XM_017011182.1 | XP_016866671.1 | 8 - 8 | 2783 | 80 | |
XM_017011184.1 | XP_016866673.1 | 8 - 8 | 2438 | 80 | |
XM_017011186.1 | XP_016866675.1 | 11 - 11 | 2671 | 80 | |
GenBank mRNA | AK000691.1 | - | 1 - 1 | 1022 | 80 |
AK172770.1 | - | 7 - 7 | 2284 | 80 | |
BC022217.1 | - | 1 - 1 | 351 | 80 | |
BC128580.1 | - | 10 - 10 | 2681 | 80 | |
BC128581.1 | - | 12 - 12 | 2401 | 80 |