The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308096.1 | NP_001295025.1 | 13 - 14 | 1304 | 128 |
NM_001324096.1 | NP_001311025.1 | 12 - 13 | 1147 | 128 | |
NM_001324097.1 | NP_001311026.1 | 12 - 13 | 1165 | 128 | |
NM_001324098.1 | NP_001311027.1 | 12 - 13 | 1357 | 128 | |
NM_032569.3 | NP_115958.2 | 13 - 14 | 1322 | 128 | |
NR_136695.1 | 14 - 15 | 1607 | 128 | ||
NR_136696.1 | 13 - 14 | 1390 | 128 | ||
NR_136697.1 | 13 - 14 | 1408 | 128 | ||
NR_136698.1 | 12 - 13 | 1339 | 128 | ||
NR_136699.1 | 12 - 13 | 1357 | 128 | ||
NR_136700.1 | 12 - 13 | 1474 | 128 | ||
XM_005255639.4 | XP_005255696.2 | 11 - 12 | 1072 | 128 | |
XM_011522716.2 | XP_011521018.1 | 13 - 14 | 1450 | 128 | |
XM_011522717.1 | XP_011521019.1 | 14 - 15 | 1507 | 128 | |
XM_017023786.1 | XP_016879275.1 | 13 - 14 | 1400 | 128 | |
XM_017023787.1 | XP_016879276.1 | 12 - 13 | 1334 | 128 | |
XM_017023788.1 | XP_016879277.1 | 14 - 15 | 1489 | 128 | |
XM_017023789.1 | XP_016879278.1 | 13 - 14 | 1456 | 128 | |
GenBank mRNA | AF244907.1 | - | 13 - 14 | 1303 | 128 |
AF326966.1 | - | 13 - 14 | 1267 | 128 | |
AK296842.1 | - | 12 - 13 | 1056 | 128 | |
AK307834.1 | - | 13 - 14 | 1297 | 128 | |
AY352585.1 | - | 13 - 14 | 1285 | 128 | |
BC003693.1 | - | 11 - 12 | 1196 | 128 | |
BC032855.1 | - | 13 - 14 | 1313 | 128 | |
BC047223.1 | - | 13 - 14 | 1302 | 128 | |
BC064940.1 | - | 14 - 15 | 1506 | 128 |