The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152401.2 | NP_689614.2 | 5 - 6 | 676 | 94 |
XM_011534374.2 | XP_011532676.1 | 5 - 6 | 655 | 94 | |
GenBank mRNA | AW629089.1 | - | 1 - 2 | 94 | |
BC034431.1 | - | 5 - 6 | 676 | 94 |