Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC2
Entrez Gene ID
160335
Gene Name
transmembrane and tetratricopeptide repeat containing 2
Gene Aliases
IBDBP1
Chromosome Location
Chr.12: 82686952 - 83134868 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320322.1 NP_001307251.1 1 - 2 518 63
NM_152588.2 NP_689801.1 1 - 2 518 63
GenBank mRNA AK315285.1 - 1 - 2 162 63
AL834509.1 - 1 - 2 518 63
AX795392.1 - 1 - 2 793 63
BC093852.1 - 1 - 2 175 63
BC093854.1 - 1 - 2 175 63
BC098572.1 - 1 - 2 672 63

View Full Product Details