Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC2
Entrez Gene ID
160335
Gene Name
transmembrane and tetratricopeptide repeat containing 2
Gene Aliases
IBDBP1
Chromosome Location
Chr.12: 82686952 - 83134868 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320322.1 NP_001307251.1 2 - 3 1084 133
NM_152588.2 NP_689801.1 2 - 3 1084 133
XM_017018884.1 XP_016874373.1 3 - 4 1431 133
GenBank mRNA AK315285.1 - 2 - 3 728 133
AL834509.1 - 2 - 3 1084 133
AX795392.1 - 2 - 3 1359 133
AX795394.1 - 3 - 4 2436 133
BC093852.1 - 2 - 3 741 133
BC093854.1 - 2 - 3 741 133
BC098572.1 - 2 - 3 1238 133

View Full Product Details