Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC2
Entrez Gene ID
160335
Gene Name
transmembrane and tetratricopeptide repeat containing 2
Gene Aliases
IBDBP1
Chromosome Location
Chr.12: 82686952 - 83134868 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320321.1 NP_001307250.1 2 - 3 1350 120
NM_001320322.1 NP_001307251.1 3 - 4 1921 120
NM_152588.2 NP_689801.1 3 - 4 1921 120
XM_017018884.1 XP_016874373.1 4 - 5 2268 120
GenBank mRNA AK074634.1 - 1 - 2 215 120
AK299771.1 - 2 - 3 1024 120
AK315285.1 - 3 - 4 1565 120
AL834509.1 - 3 - 4 1921 120
AX795392.1 - 3 - 4 2196 120
AX795394.1 - 4 - 5 3273 120
BC093852.1 - 3 - 4 1578 120
BC093854.1 - 3 - 4 1578 120
BC098572.1 - 3 - 4 2075 120
BC143378.1 - 2 - 3 1007 120

View Full Product Details