Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC2
Entrez Gene ID
160335
Gene Name
transmembrane and tetratricopeptide repeat containing 2
Gene Aliases
IBDBP1
Chromosome Location
Chr.12: 82686952 - 83134868 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320321.1 NP_001307250.1 3 - 4 1460 81
NM_001320322.1 NP_001307251.1 4 - 5 2031 81
NM_152588.2 NP_689801.1 4 - 5 2031 81
XM_017018884.1 XP_016874373.1 5 - 6 2378 81
GenBank mRNA AK074634.1 - 2 - 3 325 81
AK299771.1 - 3 - 4 1134 81
AK315285.1 - 4 - 5 1675 81
AL834509.1 - 4 - 5 2031 81
AX795392.1 - 4 - 5 2306 81
AX795394.1 - 5 - 6 3383 81
BC093852.1 - 4 - 5 1688 81
BC093854.1 - 4 - 5 1688 81
BC098572.1 - 4 - 5 2185 81
BC143378.1 - 3 - 4 1117 81
CA424966.1 - 2 - 3 81

View Full Product Details