The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152589.1 | NP_689802.1 | 12 - 13 | 1385 | 92 |
XM_017018886.1 | XP_016874375.1 | 11 - 12 | 1322 | 92 | |
XM_017018887.1 | XP_016874376.1 | 12 - 13 | 1628 | 92 | |
XM_017018888.1 | XP_016874377.1 | 10 - 11 | 1208 | 92 | |
GenBank mRNA | AK093140.1 | - | 12 - 13 | 1385 | 92 |
BC038413.2 | - | 11 - 12 | 1326 | 92 |