The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206997.1 | NP_001193926.1 | 1 - 2 | 158 | 73 |
NM_152770.2 | NP_689983.2 | 1 - 2 | 158 | 73 | |
XM_011531816.2 | XP_011530118.2 | 2 - 3 | 324 | 73 | |
XM_017007972.1 | XP_016863461.1 | 2 - 3 | 310 | 73 | |
XM_017007973.1 | XP_016863462.1 | 2 - 3 | 313 | 73 | |
XM_017007974.1 | XP_016863463.1 | 2 - 3 | 318 | 73 | |
XM_017007976.1 | XP_016863465.1 | 2 - 3 | 333 | 73 | |
XM_017007977.1 | XP_016863466.1 | 2 - 3 | 324 | 73 | |
GenBank mRNA | AK128642.1 | - | 1 - 2 | 110 | 73 |
AY349359.1 | - | 1 - 2 | 109 | 73 | |
BC034296.1 | - | 1 - 2 | 158 | 73 |