Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKKS
Entrez Gene ID
8195
Gene Name
McKusick-Kaufman syndrome
Gene Aliases
BBS6, HMCS, KMS, MKS
Chromosome Location
Chr.20: 10404780 - 10434239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018848.3 NP_061336.1 4 - 5 2056 72
NM_170784.2 NP_740754.1 4 - 5 1946 72
NR_072977.1 3 - 4 544 72
GenBank mRNA AF221992.1 - 4 - 5 1901 72
AF221993.1 - 4 - 5 2053 72
AK025741.1 - 4 - 5 1881 72
AK291925.1 - 4 - 5 1895 72
AK296439.1 - 3 - 4 538 72
AL157427.1 - 4 - 5 1886 72
BC028973.2 - 4 - 5 1849 72

View Full Product Details