The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173077.2 | NP_775100.1 | 2 - 3 | 207 | 91 |
XM_017003372.1 | XP_016858861.1 | 3 - 4 | 6314 | 91 | |
GenBank mRNA | AJ422118.1 | - | 2 - 3 | 161 | 91 |
BC112076.1 | - | 2 - 3 | 207 | 91 | |
BC112078.1 | - | 2 - 3 | 207 | 91 |