Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNC45B
Entrez Gene ID
146862
Gene Name
unc-45 myosin chaperone B
Gene Aliases
CMYA4, CTRCT43, SMUNC45, UNC-45B, UNC45
Chromosome Location
Chr.17: 35147793 - 35189345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033576.1 NP_001028748.1 2 - 3 268 83
NM_001267052.1 NP_001253981.1 2 - 3 265 83
NM_001308281.1 NP_001295210.1 1 - 2 171 83
NM_173167.3 NP_775259.1 1 - 2 171 83
XM_017024234.1 XP_016879723.1 2 - 3 289 83
GenBank mRNA AF539794.1 - 1 - 2 171 83
AL832355.1 - 2 - 3 262 83
AL833281.1 - 2 - 3 241 83
BC101062.2 - 1 - 2 171 83
BC101063.2 - 1 - 2 83
BX647642.1 - 2 - 3 268 83

View Full Product Details