The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173481.3 | NP_775752.1 | 4 - 5 | 2063 | 90 |
NR_135168.1 | 3 - 4 | 226 | 90 | ||
XM_011527685.2 | XP_011525987.1 | 4 - 5 | 2108 | 90 | |
XM_011527686.2 | XP_011525988.1 | 4 - 5 | 2201 | 90 | |
GenBank mRNA | BC042125.2 | - | 4 - 5 | 2030 | 90 |
BC052236.1 | - | 4 - 5 | 2050 | 90 | |
BX648389.1 | - | 4 - 5 | 2044 | 90 | |
BX648815.1 | - | 3 - 4 | 1251 | 90 | |
HY173833.1 | - | 3 - 4 | 212 | 90 |