Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUT11
Entrez Gene ID
170384
Gene Name
fucosyltransferase 11
Gene Aliases
-
Chromosome Location
Chr.10: 73772175 - 73780251 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173540.2 NP_775811.2 2 - 3 1377 69
GenBank mRNA AK314944.1 - 2 - 3 1359 69
BC036037.1 - 2 - 3 1377 69
BC100996.1 - 2 - 3 1374 69
BC100997.1 - 2 - 3 1374 69

View Full Product Details