The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173560.3 | NP_775831.2 | 15 - 16 | 1738 | 110 |
XM_011535589.1 | XP_011533891.1 | 14 - 15 | 1625 | 110 | |
XM_017010477.1 | XP_016865966.1 | 14 - 15 | 1545 | 110 | |
GenBank mRNA | BC039248.1 | - | 15 - 16 | 1738 | 110 |