The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173573.2 | NP_775844.2 | 7 - 8 | 775 | 84 |
XM_011519965.2 | XP_011518267.2 | 7 - 8 | 950 | 84 | |
XM_011519967.2 | XP_011518269.2 | 7 - 8 | 950 | 84 | |
XM_017017476.1 | XP_016872965.1 | 7 - 8 | 950 | 84 | |
XM_017017477.1 | XP_016872966.1 | 7 - 8 | 935 | 84 | |
XM_017017478.1 | XP_016872967.1 | 7 - 8 | 791 | 84 | |
XM_017017479.1 | XP_016872968.1 | 7 - 8 | 950 | 84 | |
GenBank mRNA | BC039077.1 | - | 7 - 8 | 775 | 84 |