Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGMB
Entrez Gene ID
285704
Gene Name
repulsive guidance molecule family member b
Gene Aliases
DRAGON
Chromosome Location
Chr.5: 98768386 - 98796494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017009390.1 XP_016864879.1 1 - 2 335 72
GenBank mRNA AK074887.1 - 1 - 2 334 72

View Full Product Details