Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location
Chr.13: 20566446 - 20691437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318493.1 NP_001305422.1 4 - 5 927 74
NM_006531.4 NP_006522.2 3 - 4 599 74
NM_175605.4 NP_783195.2 5 - 6 828 74
NR_134653.1 4 - 5 927 74
XM_005266553.2 XP_005266610.1 5 - 6 1189 74
XM_006719870.3 XP_006719933.1 3 - 4 294 74
XM_011535241.2 XP_011533543.1 4 - 5 402 74
XM_011535242.1 XP_011533544.1 6 - 7 1329 74
XM_011535243.1 XP_011533545.1 5 - 6 1208 74
XM_017020757.1 XP_016876246.1 4 - 5 1307 74
XM_017020759.1 XP_016876248.1 4 - 5 860 74
XM_017020760.1 XP_016876249.1 5 - 6 761 74
XM_017020761.1 XP_016876250.1 4 - 5 397 74
XM_017020762.1 XP_016876251.1 4 - 5 860 74
XM_017020763.1 XP_016876252.1 4 - 5 860 74
XM_017020764.1 XP_016876253.1 3 - 4 306 74
XM_017020765.1 XP_016876254.1 3 - 4 306 74
XM_017020768.1 XP_016876257.1 4 - 5 860 74
XM_017020770.1 XP_016876259.1 5 - 6 761 74
XM_017020771.1 XP_016876260.1 5 - 6 1189 74
XM_017020772.1 XP_016876261.1 3 - 4 306 74
XM_017020774.1 XP_016876263.1 3 - 4 252 74
XM_017020775.1 XP_016876264.1 4 - 5 860 74
XM_017020776.1 XP_016876265.1 4 - 5 860 74
GenBank mRNA AK126668.1 - 4 - 5 1298 74
AK303853.1 - 3 - 4 253 74
AK310223.1 - 5 - 6 484 74
BC030776.1 - 5 - 6 509 74
U20362.1 - 3 - 4 343 74

View Full Product Details