Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location
Chr.13: 20566446 - 20691437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318491.1 NP_001305420.1 7 - 8 882 94
NM_001318493.1 NP_001305422.1 9 - 10 1267 94
NM_006531.4 NP_006522.2 8 - 9 939 94
NM_175605.4 NP_783195.2 10 - 11 1168 94
NR_134653.1 9 - 10 1267 94
XM_005266553.2 XP_005266610.1 10 - 11 1529 94
XM_006719870.3 XP_006719933.1 8 - 9 634 94
XM_011535241.2 XP_011533543.1 9 - 10 742 94
XM_011535242.1 XP_011533544.1 11 - 12 1669 94
XM_011535243.1 XP_011533545.1 10 - 11 1548 94
XM_017020757.1 XP_016876246.1 9 - 10 1647 94
XM_017020758.1 XP_016876247.1 8 - 9 1143 94
XM_017020759.1 XP_016876248.1 9 - 10 1200 94
XM_017020760.1 XP_016876249.1 10 - 11 1101 94
XM_017020761.1 XP_016876250.1 9 - 10 737 94
XM_017020762.1 XP_016876251.1 9 - 10 1200 94
XM_017020763.1 XP_016876252.1 9 - 10 1200 94
XM_017020764.1 XP_016876253.1 8 - 9 646 94
XM_017020765.1 XP_016876254.1 8 - 9 646 94
XM_017020766.1 XP_016876255.1 7 - 8 557 94
XM_017020767.1 XP_016876256.1 7 - 8 568 94
XM_017020768.1 XP_016876257.1 9 - 10 1200 94
XM_017020769.1 XP_016876258.1 7 - 8 569 94
XM_017020770.1 XP_016876259.1 10 - 11 1101 94
XM_017020771.1 XP_016876260.1 10 - 11 1529 94
XM_017020772.1 XP_016876261.1 8 - 9 646 94
XM_017020773.1 XP_016876262.1 9 - 10 1711 94
XM_017020774.1 XP_016876263.1 7 - 8 522 94
XM_017020775.1 XP_016876264.1 9 - 10 1200 94
XM_017020776.1 XP_016876265.1 9 - 10 1200 94
GenBank mRNA AK126668.1 - 9 - 10 1638 94
AK300769.1 - 7 - 8 546 94
AK303853.1 - 7 - 8 523 94
AK308281.1 - 8 - 9 594 94
AK310223.1 - 10 - 11 824 94
BC030776.1 - 10 - 11 849 94
U20362.1 - 8 - 9 683 94

View Full Product Details