Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
Chromosome Location
Chr.13: 20566446 - 20691437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318491.1 NP_001305420.1 8 - 9 991 114
NM_001318493.1 NP_001305422.1 10 - 11 1376 114
NM_006531.4 NP_006522.2 9 - 10 1048 114
NM_175605.4 NP_783195.2 11 - 12 1277 114
NR_134653.1 10 - 11 1376 114
XM_005266553.2 XP_005266610.1 11 - 12 1638 114
XM_006719870.3 XP_006719933.1 9 - 10 743 114
XM_011535241.2 XP_011533543.1 10 - 11 851 114
XM_011535242.1 XP_011533544.1 12 - 13 1778 114
XM_011535243.1 XP_011533545.1 11 - 12 1657 114
XM_017020757.1 XP_016876246.1 10 - 11 1756 114
XM_017020758.1 XP_016876247.1 9 - 10 1252 114
XM_017020759.1 XP_016876248.1 10 - 11 1309 114
XM_017020760.1 XP_016876249.1 11 - 12 1210 114
XM_017020761.1 XP_016876250.1 10 - 11 846 114
XM_017020762.1 XP_016876251.1 10 - 11 1309 114
XM_017020763.1 XP_016876252.1 10 - 11 1309 114
XM_017020764.1 XP_016876253.1 9 - 10 755 114
XM_017020765.1 XP_016876254.1 9 - 10 755 114
XM_017020766.1 XP_016876255.1 8 - 9 666 114
XM_017020767.1 XP_016876256.1 8 - 9 677 114
XM_017020768.1 XP_016876257.1 10 - 11 1309 114
XM_017020769.1 XP_016876258.1 8 - 9 678 114
XM_017020770.1 XP_016876259.1 11 - 12 1210 114
XM_017020771.1 XP_016876260.1 11 - 12 1638 114
XM_017020772.1 XP_016876261.1 9 - 10 755 114
XM_017020773.1 XP_016876262.1 10 - 11 1820 114
XM_017020774.1 XP_016876263.1 8 - 9 631 114
XM_017020775.1 XP_016876264.1 10 - 11 1309 114
XM_017020776.1 XP_016876265.1 10 - 11 1309 114
GenBank mRNA AK126668.1 - 10 - 11 1747 114
AK300769.1 - 8 - 9 655 114
AK303853.1 - 8 - 9 632 114
AK308281.1 - 9 - 10 703 114
BC030776.1 - 11 - 12 958 114
BX640833.1 - 1 - 2 126 114

View Full Product Details