The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173207.2 | NP_775299.1 | 1 - 2 | 206 | 61 |
GenBank mRNA | AK291112.1 | - | 1 - 2 | 162 | 61 |
BU657748.1 | - | 1 - 2 | 182 | 61 |